Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159860.1 | NP_001153332.1 | 5 | 674 | ||
NM_001159861.1 | NP_001153333.1 | 5 | 679 | |||
NM_152478.2 | NP_689691.2 | 5 | 726 | |||
XM_005258558.1 | XP_005258615.1 | 3 | 512 | |||
XM_011526517.2 | XP_011524819.1 | 5 | 2510 | |||
XM_017026346.1 | XP_016881835.1 | 5 | 825 | |||
XM_017026347.1 | XP_016881836.1 | 5 | 802 | |||
XM_017026348.1 | XP_016881837.1 | 5 | 2409 | |||
XM_017026349.1 | XP_016881838.1 | 5 | 622 | |||
XM_017026350.1 | XP_016881839.1 | 5 | 655 | |||
XM_017026351.1 | XP_016881840.1 | 6 | 924 | |||
XM_017026352.1 | XP_016881841.1 | 6 | 901 | |||
XM_017026353.1 | XP_016881842.1 | 6 | 2683 | |||
GenBank mRNA | AK055592.1 | - | 5 | 702 | ||
AL833236.1 | - | 1 | 18 | |||
BC111772.1 | - | 4 | 608 | 40034375 |