Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174905.3 | NP_777565.3 | 8 | 1223 | ||
GenBank mRNA | AK128745.1 | - | 5 | 4089 | ||
BC034383.1 | - | 6 | 761 | |||
BC080606.1 | - | 6 | 921 | |||
BI871449.1 | - | 5 | 756 |