Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174905.3 | NP_777565.3 | 3, 4 | 372 | ||
XM_017026354.1 | XP_016881843.1 | 3, 4 | 383 | |||
GenBank mRNA | AK128745.1 | - | 2, 3 | 290 | ||
BC080606.1 | - | 3, 4 | 316 | |||
BC117258.1 | - | 3, 4 | 372 | 40125809 | ||
BC143830.1 | - | 3, 4 | 372 | 9052345 | ||
BM561971.1 | - | 3, 4 | 389 |