Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 17, 18 | 2030 | ||
NM_207325.2 | NP_997208.2 | 17, 18 | 2056 | |||
XM_006723045.1 | XP_006723108.1 | 14, 15 | 1572 | |||
XM_011526526.2 | XP_011524828.1 | 17, 18 | 2011 | |||
XM_017026366.1 | XP_016881855.1 | 16, 17 | 1945 | |||
XM_017026367.1 | XP_016881856.1 | 17, 18 | 1970 | |||
GenBank mRNA | AK125846.1 | - | 1, 2 | 174 | ||
AK126264.1 | - | 12, 13 | 1284 | |||
AK126757.1 | - | 17, 18 | 2047 | |||
AK129497.1 | - | 17, 18 | 2011 | |||
AL833713.1 | - | 7, 8 | 2009 | |||
BC131725.1 | - | 16, 17 | 1878 | 40123868 | ||
CR749459.1 | - | 11, 12 | 1226 |