Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 15 | 1776 | ||
NM_207325.2 | NP_997208.2 | 15 | 1802 | |||
XM_006723045.1 | XP_006723108.1 | 12 | 1318 | |||
XM_011526526.2 | XP_011524828.1 | 15 | 1757 | |||
XM_017026366.1 | XP_016881855.1 | 14 | 1691 | |||
XM_017026367.1 | XP_016881856.1 | 15 | 1716 | |||
GenBank mRNA | AK126264.1 | - | 9 | 877 | ||
AK126757.1 | - | 15 | 1793 | |||
AK129497.1 | - | 15 | 1757 | |||
AL833713.1 | - | 5 | 498 | |||
BC131725.1 | - | 14 | 1624 | 40123868 | ||
CR749459.1 | - | 9 | 972 |