Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178002.2 | NP_001171473.1 | 13 | 1445 | ||
NM_138813.3 | NP_620168.1 | 13 | 1566 | |||
NR_047593.2 | 13 | 1793 | ||||
XM_006722654.2 | XP_006722717.1 | 13 | 1566 | |||
XM_006722655.2 | XP_006722718.1 | 13 | 1645 | |||
XM_006722656.2 | XP_006722719.1 | 13 | 1454 | |||
XM_011527707.1 | XP_011526009.1 | 13 | 1566 | |||
XM_011527708.1 | XP_011526010.1 | 13 | 1566 | |||
XM_011527709.1 | XP_011526011.1 | 13 | 1566 | |||
XM_011527710.2 | XP_011526012.1 | 7 | 775 | |||
XM_011527711.2 | XP_011526013.1 | 13 | 1566 | |||
GenBank mRNA | AK125225.1 | - | 13 | 1348 | ||
AK125969.1 | - | 13 | 1793 | |||
AY302538.1 | - | 13 | 1408 | |||
BC035162.2 | - | 13 | 1561 | 5265541 |