Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152484.2 | NP_689697.2 | 6 | 1089 | ||
XM_006723046.2 | XP_006723109.1 | 8 | 1416 | |||
XM_006723047.3 | XP_006723110.1 | 7 | 1339 | |||
XM_006723048.3 | XP_006723111.1 | 7 | 1263 | |||
XM_011526538.2 | XP_011524840.1 | 7 | 1178 | |||
XM_011526539.2 | XP_011524841.1 | 5 | 672 | |||
XM_017026376.1 | XP_016881865.1 | 6 | 924 | |||
XM_017026377.1 | XP_016881866.1 | 6 | 1003 | |||
XM_017026378.1 | XP_016881867.1 | 7 | 1041 | |||
XM_017026379.1 | XP_016881868.1 | 4 | 890 | |||
XM_017026380.1 | XP_016881869.1 | 4 | 698 | |||
XM_017026381.1 | XP_016881870.1 | 5 | 1101 | |||
XM_017026382.1 | XP_016881871.1 | 4 | 798 | |||
GenBank mRNA | AK122846.1 | - | 3 | 410 | ||
AK289987.1 | - | 6 | 862 | |||
AL833408.1 | - | 6 | 1090 | |||
AY835704.1 | - | 6 | 1089 | |||
BC059407.1 | - | 5 | 718 | |||
BC117219.1 | - | 2 | 346 | 40125770 |