Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001406.3 | NP_001397.1 | 5 | 2367 | ||
GenBank mRNA | AK027329.1 | - | 2 | 1490 | ||
BC022499.1 | - | 4 | 1806 | |||
U66406.1 | - | 5 | 2583 |