Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM163A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173509.2 | NP_775780.1 | 5 | 520 | ||
XM_005244913.2 | XP_005244970.1 | 6 | 957 | |||
XM_006711174.2 | XP_006711237.1 | 6 | 668 | |||
XM_006711175.2 | XP_006711238.1 | 7 | 780 | |||
XM_006711176.2 | XP_006711239.1 | 5 | 646 | |||
XM_006711177.1 | XP_006711240.1 | 6 | 500 | |||
XM_011509217.1 | XP_011507519.1 | 8 | 802 | |||
XM_011509221.1 | XP_011507523.1 | 7 | 608 | |||
XM_011509226.1 | XP_011507528.1 | 5 | 442 | |||
XM_011509228.1 | XP_011507530.1 | 5 | 576 | |||
XM_017000386.1 | XP_016855875.1 | 8 | 951 | |||
XM_017000387.1 | XP_016855876.1 | 9 | 1073 | |||
XM_017000388.1 | XP_016855877.1 | 7 | 814 | |||
XM_017000389.1 | XP_016855878.1 | 9 | 1085 | |||
XM_017000390.1 | XP_016855879.1 | 7 | 805 | |||
XM_017000391.1 | XP_016855880.1 | 7 | 929 | |||
XM_017000392.1 | XP_016855881.1 | 6 | 454 | |||
XM_017000393.1 | XP_016855882.1 | 5 | 313 | |||
GenBank mRNA | AB299025.1 | - | 2 | 124 | ||
AK292432.1 | - | 6 | 584 | |||
AK310711.1 | - | 5 | 576 | |||
AK313887.1 | - | 5 | 327 | |||
AY902195.1 | - | 5 | 520 | |||
BC009382.2 | - | 5 | 520 | 4124490 |