Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001407.2 | NP_001398.2 | 33 | 9222 | ||
GenBank mRNA | AB011536.1 | - | 23 | 3098 | ||
AF231023.1 | - | 33 | 9222 | |||
AK299868.1 | - | 4 | 458 |
Set Membership: |
G-protein coupled receptor |