Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF436-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033690.1 | 1 | 2666 | |||
NR_033691.1 | 3 | 491 | ||||
GenBank mRNA | AK074989.1 | - | 1 | 2581 | ||
BC028411.1 | - | 1 | 2407 | 4822173 | ||
BC054115.1 | - | 1 | 2670 | 5270027 | ||
BC068594.1 | - | 1 | 2648 | 4828713 | ||
BX098506.1 | - | 3 | 459 |