Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322302.1 | NP_001309231.1 | 3 | 383 | ||
NM_001322303.1 | NP_001309232.1 | 2 | 317 | |||
NM_001322304.1 | NP_001309233.1 | 3 | 461 | |||
NM_001322305.1 | NP_001309234.1 | 2 | 803 | |||
NM_152495.1 | NP_689708.1 | 2 | 803 | |||
NR_136287.1 | 2 | 803 | ||||
NR_136288.1 | 3 | 313 | ||||
NR_136289.1 | 4 | 449 | ||||
NR_136290.1 | 3 | 383 | ||||
NR_136291.1 | 4 | 519 | ||||
NR_136292.1 | 3 | 453 | ||||
NR_136293.1 | 6 | 689 | ||||
NR_136294.1 | 5 | 623 | ||||
NR_136295.1 | 4 | 494 | ||||
NR_136296.1 | 4 | 519 | ||||
NR_136297.1 | 3 | 383 | ||||
GenBank mRNA | AK096312.1 | - | 2 | 803 | ||
BC022780.1 | - | 2 | 291 | 3894832 |