Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198149.2 | NP_937792.2 | 5 | 1090 | ||
NR_030775.1 | 5 | 812 | ||||
XM_017000419.1 | XP_016855908.1 | 5 | 829 | |||
GenBank mRNA | AK294106.1 | - | 5 | 846 | ||
AK316312.1 | - | 5 | 1088 | |||
AY358589.1 | - | 5 | 863 | |||
BC009558.1 | - | 5 | 867 | |||
BC061908.1 | - | 4 | 709 | |||
BC104444.1 | - | 5 | 824 | 40022864 | ||
BC104445.1 | - | 5 | 824 | 40022873 | ||
BC111855.1 | - | 5 | 1078 | 5762600 | ||
BM923401.1 | - | 813 |