Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018060.2 | NP_001018070.1 | 13 | 1389 | ||
NM_001146288.1 | NP_001139760.1 | 13 | 1474 | |||
NM_144704.2 | NP_653305.1 | 13 | 1389 | |||
NR_027464.1 | 12 | 1346 | ||||
GenBank mRNA | AK055035.1 | - | 13 | 1373 | ||
AK094844.1 | - | 13 | 1378 | |||
AK126917.1 | - | 1 | 19 | |||
AK295581.1 | - | 12 | 1249 | |||
AK316042.1 | - | 12 | 1242 | |||
AY927574.1 | - | 1 | 819 | |||
BC032485.1 | - | 13 | 1356 | 5221203 | ||
CR456342.1 | - | 12 | 1208 |