Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529914.2 | XP_011528216.1 | 3 | 593 | ||
XM_011529917.2 | XP_011528219.1 | 3 | 452 | |||
XM_011529918.1 | XP_011528220.1 | 3 | 593 | |||
XM_017028621.1 | XP_016884110.1 | 3 | 474 | |||
XM_017028622.1 | XP_016884111.1 | 3 | 393 | |||
XM_017028624.1 | XP_016884113.1 | 3 | 593 | |||
XM_017028625.1 | XP_016884114.1 | 1 | 29 | |||
GenBank mRNA | AK098703.1 | - | 3 | 418 | ||
BC030013.1 | - | 3 | 203 | 4823215 | ||
BI459310.1 | - | 3 | 198 | |||
CR456341.1 | - | 2 | 180 | |||
HY002382.1 | - | 4 | 243 |