Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152512.3 | NP_689725.2 | 2 | 580 | ||
XM_006724149.3 | XP_006724212.1 | 2 | 1175 | |||
XM_006724150.3 | XP_006724213.1 | 2 | 365 | |||
XM_006724153.3 | XP_006724216.1 | 2 | 674 | |||
XM_011529927.2 | XP_011528229.1 | 2 | 674 | |||
XM_011529928.2 | XP_011528230.1 | 2 | 674 | |||
XM_011529931.2 | XP_011528233.1 | 2 | 674 | |||
GenBank mRNA | BC033895.2 | - | 2 | 580 | 4831324 | |
CR456452.1 | - | 1 | 375 |