Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207327.4 | NP_997210.3 | 4 | 353 | ||
XM_011529960.1 | XP_011528262.1 | 4 | 740 | |||
XM_011529962.2 | XP_011528264.1 | 4 | 348 | |||
XM_011529965.2 | XP_011528267.1 | 4 | 669 | |||
GenBank mRNA | AK127072.1 | - | 1 | 573 | ||
BC036462.1 | - | 5 | 474 | |||
BC067871.1 | - | 4 | 353 | 30338205 | ||
BX648331.1 | - | 4 | 361 | |||
CR456415.1 | - | 2 | 163 |