Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153712.4 | NP_714923.1 | 5 | 982 | ||
XM_005263599.3 | XP_005263656.1 | 5 | 1021 | |||
XM_011510665.2 | XP_011508967.1 | 5 | 1021 | |||
GenBank mRNA | AB071393.2 | - | 5 | 827 | ||
AK126549.1 | - | 3 | 474 | |||
BC036819.1 | - | 5 | 994 | 5767758 | ||
BX538316.1 | - | 1 | 193 | |||
BX647135.1 | - | 2 | 280 |