Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015880.1 | NP_001015880.1 | 6 | 921 | ||
NM_004670.3 | NP_004661.2 | 6 | 921 | |||
GenBank mRNA | AF074331.1 | - | 6 | 721 | ||
AF091242.1 | - | 6 | 738 | |||
AF150754.2 | - | 6 | 738 | |||
AF173365.1 | - | 6 | 706 | |||
AF313907.1 | - | 6 | 658 | |||
AK301453.1 | - | 6 | 708 | |||
AK309262.1 | - | 6 | 730 | |||
AK312556.1 | - | 6 | 762 | |||
BC009894.2 | - | 6 | 769 | 2988345 |