Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198274.3 | NP_938015.1 | 4 | 720 | ||
XM_005264156.2 | XP_005264213.1 | 4 | 675 | |||
GenBank mRNA | AK091724.1 | - | 4 | 638 | ||
AL832035.1 | - | 4 | 637 | |||
AY518933.1 | - | 4 | 617 | |||
AY518934.1 | - | 1 | 25 | |||
BC126191.1 | - | 4 | 720 | 8991907 | ||
BX647878.1 | - | 4 | 639 |