Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165977.2 | NP_001159449.1 | 14, 15 | 1858 | ||
NM_001165978.2 | NP_001159450.1 | 14, 15 | 1858 | |||
NM_001321070.1 | NP_001307999.1 | 14, 15 | 1849 | |||
NM_144707.3 | NP_653308.2 | 14, 15 | 1858 | |||
XM_006712301.2 | XP_006712364.1 | 11, 12 | 1118 | |||
GenBank mRNA | AF245303.1 | - | 14, 15 | 1853 | ||
AF245304.1 | - | 14, 15 | 1853 | |||
AK091175.1 | - | 14, 15 | 1828 | |||
AK290366.1 | - | 14, 15 | 1813 | |||
AK313267.1 | - | 14, 15 | 1814 | |||
AY358377.1 | - | 14, 15 | 1809 | |||
BC113877.1 | - | 10, 11 | 2525 | |||
BC114525.1 | - | 14, 15 | 1833 | 40054105 |