Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144708.3 | NP_653309.3 | 21 | 3994 | ||
XM_011510673.2 | XP_011508975.1 | 21 | 4513 | |||
XM_011510674.1 | XP_011508976.1 | 21 | 4480 | |||
XM_011510675.2 | XP_011508977.1 | 20 | 4408 | |||
XM_011510676.2 | XP_011508978.1 | 21 | 4513 | |||
XM_011510677.2 | XP_011508979.1 | 21 | 4513 | |||
XM_011510679.2 | XP_011508981.1 | 19 | 4084 | |||
XM_011510685.2 | XP_011508987.1 | 21 | 4069 | |||
XM_011510686.2 | XP_011508988.1 | 17 | 4006 | |||
XM_011510687.2 | XP_011508989.1 | 20 | 3877 | |||
XM_011510691.2 | XP_011508993.1 | 17 | 2699 | |||
XM_011510692.2 | XP_011508994.1 | 19 | 3713 | |||
XM_011510693.2 | XP_011508995.1 | 18 | 3609 | |||
XM_011510694.2 | XP_011508996.1 | 17 | 3428 | |||
XM_017003415.1 | XP_016858904.1 | 16 | 3208 | |||
XM_017003416.1 | XP_016858905.1 | 18 | 3620 | |||
XM_017003418.1 | XP_016858907.1 | 16 | 3323 | |||
XM_017003419.1 | XP_016858908.1 | 16 | 3290 | |||
XM_017003420.1 | XP_016858909.1 | 16 | 3226 | |||
GenBank mRNA | AJ549812.1 | - | 20 | 3693 | ||
AK058144.1 | - | 9 | 1346 | |||
BC044907.1 | - | 17 | 3487 | 4836761 | ||
BC047413.1 | - | 24 | 3558 |