Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080410.2 | NP_001073879.2 | 10, 11 | 2458 | ||
XM_005264158.3 | XP_005264215.1 | 8, 9 | 1377 | |||
XM_006711950.2 | XP_006712013.1 | 15, 16 | 3442 | |||
XM_011532562.1 | XP_011530864.1 | 11, 12 | 2890 | |||
XM_011532563.2 | XP_011530865.1 | 9, 10 | 1815 | |||
XM_017003421.1 | XP_016858910.1 | 12, 13 | 5539 | |||
XM_017003422.1 | XP_016858911.1 | 15, 16 | 5184 | |||
XM_017003423.1 | XP_016858912.1 | 15, 16 | 4565 | |||
GenBank mRNA | AB075820.1 | - | 10, 11 | 2296 | ||
AK095028.1 | - | 1, 2 | 129 | |||
AK127503.1 | - | 7, 8 | 939 | |||
BC112202.1 | - | 9, 10 | 1366 |