Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM84A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145175.2 | NP_660158.2 | 2 | 1044 | ||
GenBank mRNA | AF370404.1 | - | 2 | 986 | ||
AJ417080.1 | - | 2 | 998 | |||
AK092654.1 | - | 3 | 844 | |||
AK314463.1 | - | 2 | 1013 | |||
BC026346.1 | - | 1 | 1038 | 4812838 | ||
BC029933.1 | - | 2 | 998 | |||
BC052284.1 | - | 2 | 993 | 5454643 |