Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDSUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128212.2 | NP_001121684.1 | 2 | 395 | ||
NM_001128213.1 | NP_001121685.1 | 2 | 411 | |||
NM_001307994.1 | NP_001294923.1 | 2 | 395 | |||
NM_152528.2 | NP_689741.2 | 2 | 303 | |||
XM_005246355.2 | XP_005246412.2 | 2 | 389 | |||
XM_011510753.2 | XP_011509055.1 | 2 | 355 | |||
XM_017003481.1 | XP_016858970.1 | 2 | 389 | |||
XM_017003482.1 | XP_016858971.1 | 2 | 389 | |||
XM_017003483.1 | XP_016858972.1 | 2 | 304 | |||
GenBank mRNA | AK093494.1 | - | 2 | 303 | ||
AK129983.1 | - | 2 | 411 | |||
BC029520.1 | - | 2 | 299 | 5301559 | ||
DR000438.1 | - | 2 | 402 |