Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138287.3 | NP_612144.1 | 4 | 2175 | ||
GenBank mRNA | AF484416.1 | - | 4 | 2086 | ||
AK125086.1 | - | 4 | 2175 | |||
AL833598.1 | - | 4 | 2108 | |||
AY225123.1 | - | 4 | 2220 | |||
AY780792.1 | - | 3 | 550 | |||
BC042191.1 | - | 4 | 2094 | 4339554 | ||
BC060509.1 | - | 4 | 2117 | 6457163 | ||
BX648267.1 | - | 1 | 2030 | |||
BX648645.1 | - | 4 | 1928 |