Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001175.6 | NP_001166.3 | 3, 4 | 505 | ||
NM_001321420.1 | NP_001308349.1 | 4, 5 | 663 | |||
NM_001321421.1 | NP_001308350.1 | 3, 4 | 384 | |||
NM_001321422.1 | NP_001308351.1 | 3, 4 | 506 | |||
NM_001321423.1 | NP_001308352.1 | 3, 4 | 372 | |||
GenBank mRNA | AB451315.1 | - | 2, 3 | 259 | ||
AB451445.1 | - | 2, 3 | 259 | |||
AF498927.1 | - | 2, 3 | 259 | |||
AK311807.1 | - | 3, 4 | 335 | |||
AV685495.1 | - | 3, 4 | 364 | |||
AV717900.1 | - | 3, 4 | 383 | |||
BC009200.2 | - | 3, 4 | 309 | 3621138 | ||
BE744996.1 | - | 3, 4 | 370 | |||
BG390701.1 | - | 4, 5 | 480 | |||
BI762648.1 | - | 3, 4 | 504 | |||
BP355037.1 | - | 3, 4 | 480 | |||
BY996333.1 | - | 3, 4 | 369 | |||
CD385175.1 | - | 3, 4 | 523 | |||
L07916.1 | - | 3, 4 | 333 | |||
L20688.1 | - | 3, 4 | 411 | |||
X69549.1 | - | 3, 4 | 313 |