Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MB21D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178496.3 | NP_848591.2 | 2 | 823 | ||
GenBank mRNA | AK056276.1 | - | 1 | 219 | ||
AK290447.1 | - | 2 | 518 | |||
BC013123.1 | - | 2 | 501 | 4471808 | ||
BC036194.1 | - | 2 | 807 | 5267040 | ||
BC045582.1 | - | 2 | 823 | 5262015 | ||
BX648517.1 | - | 2 | 522 |