Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144718.3 | NP_653319.1 | 14 | 2047 | ||
XM_011512461.2 | XP_011510763.1 | 14 | 1804 | |||
XM_017005775.1 | XP_016861264.1 | 14 | 1804 | |||
XM_017005776.1 | XP_016861265.1 | 14 | 1941 | |||
XM_017005777.1 | XP_016861266.1 | 14 | 1864 | |||
XM_017005778.1 | XP_016861267.1 | 14 | 2048 | |||
GenBank mRNA | AK126897.1 | - | 10 | 1453 | ||
AK129575.1 | - | 4 | 576 | |||
AL832148.1 | - | 14 | 2016 | |||
AL832327.2 | - | 14 | 3712 | |||
AY099107.1 | - | 14 | 1967 | |||
BC036951.1 | - | 14 | 1988 | 5500345 | ||
BC038595.1 | - | 2 | 170 |