Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320276.1 | NP_001307205.1 | 7, 8 | 3272 | ||
NM_152536.3 | NP_689749.3 | 7, 8 | 3272 | |||
XM_011533422.1 | XP_011531724.1 | 7, 8 | 3272 | |||
XM_011533423.1 | XP_011531725.1 | 7, 8 | 3272 | |||
GenBank mRNA | AK097276.1 | - | 7, 8 | 1785 | ||
AK123054.1 | - | 4, 5 | 464 | |||
AK131078.1 | - | 7, 8 | 2936 | |||
BC035364.1 | - | 4, 5 | 709 | |||
BC132814.1 | - | 7, 8 | 2910 | 40146836 | ||
BC132816.1 | - | 7, 8 | 2910 | 40146838 | ||
BC144300.1 | - | 7, 8 | 2910 | 9052820 | ||
BX538312.1 | - | 7, 8 | 1413 | |||
BX640820.1 | - | 7, 8 | 2213 |