Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015887.1 | NP_001015887.1 | 3 | 562 | ||
NM_152538.2 | NP_689751.2 | 5 | 693 | |||
XM_006713517.1 | XP_006713580.1 | 4 | 613 | |||
XM_011512462.2 | XP_011510764.1 | 4 | 771 | |||
XM_011512463.1 | XP_011510765.1 | 5 | 516 | |||
XM_011512464.2 | XP_011510766.1 | 5 | 504 | |||
XM_011512465.1 | XP_011510767.1 | 5 | 558 | |||
XM_011512466.2 | XP_011510768.1 | 4 | 775 | |||
XM_011512467.2 | XP_011510769.1 | 4 | 774 | |||
XM_011512468.1 | XP_011510770.1 | 6 | 706 | |||
XM_011512469.1 | XP_011510771.1 | 4 | 366 | |||
XM_011512470.1 | XP_011510772.1 | 4 | 581 | |||
XM_011512471.2 | XP_011510773.1 | 4 | 775 | |||
XM_011512472.1 | XP_011510774.1 | 4 | 454 | |||
XM_011512474.2 | XP_011510776.1 | 4 | 784 | |||
XM_017005789.1 | XP_016861278.1 | 3 | 351 | |||
XM_017005790.1 | XP_016861279.1 | 4 | 613 | |||
GenBank mRNA | AB071618.1 | - | 3 | 552 | ||
AB079879.1 | - | 3 | 315 | |||
AK097667.1 | - | 5 | 677 | |||
AK292387.1 | - | 4 | 581 | |||
AK296503.1 | - | 3 | 461 | |||
AY358141.1 | - | 3 | 603 | |||
BC034411.1 | - | 5 | 486 | 5172176 |