Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015887.1 | NP_001015887.1 | 4 | 764 | ||
NM_152538.2 | NP_689751.2 | 6 | 895 | |||
XM_006713517.1 | XP_006713580.1 | 5 | 815 | |||
XM_011512462.2 | XP_011510764.1 | 5 | 973 | |||
XM_011512463.1 | XP_011510765.1 | 6 | 718 | |||
XM_011512464.2 | XP_011510766.1 | 6 | 706 | |||
XM_011512465.1 | XP_011510767.1 | 6 | 760 | |||
XM_011512466.2 | XP_011510768.1 | 5 | 977 | |||
XM_011512467.2 | XP_011510769.1 | 5 | 976 | |||
XM_011512468.1 | XP_011510770.1 | 7 | 908 | |||
XM_011512469.1 | XP_011510771.1 | 5 | 568 | |||
XM_011512470.1 | XP_011510772.1 | 5 | 783 | |||
XM_011512471.2 | XP_011510773.1 | 5 | 977 | |||
XM_011512472.1 | XP_011510774.1 | 5 | 656 | |||
XM_011512474.2 | XP_011510776.1 | 5 | 986 | |||
XM_017005789.1 | XP_016861278.1 | 4 | 553 | |||
XM_017005790.1 | XP_016861279.1 | 5 | 815 | |||
GenBank mRNA | AB071618.1 | - | 4 | 754 | ||
AB079879.1 | - | 4 | 517 | |||
AK097667.1 | - | 6 | 879 | |||
AK122611.1 | - | 4 | 727 | |||
AK292387.1 | - | 5 | 783 | |||
AK296503.1 | - | 4 | 663 | |||
AL834256.1 | - | 2 | 195 | |||
AY358141.1 | - | 4 | 805 | |||
BC034411.1 | - | 6 | 688 | 5172176 |