Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015887.1 | NP_001015887.1 | 7 | 1488 | ||
NM_152538.2 | NP_689751.2 | 9 | 1619 | |||
XM_006713517.1 | XP_006713580.1 | 8 | 1539 | |||
XM_011512462.2 | XP_011510764.1 | 8 | 1697 | |||
XM_011512463.1 | XP_011510765.1 | 9 | 1442 | |||
XM_011512464.2 | XP_011510766.1 | 9 | 1430 | |||
XM_011512465.1 | XP_011510767.1 | 9 | 1484 | |||
XM_011512466.2 | XP_011510768.1 | 8 | 1629 | |||
XM_011512467.2 | XP_011510769.1 | 8 | 1616 | |||
XM_011512468.1 | XP_011510770.1 | 10 | 1632 | |||
XM_011512469.1 | XP_011510771.1 | 8 | 1292 | |||
XM_011512470.1 | XP_011510772.1 | 8 | 1507 | |||
XM_011512471.2 | XP_011510773.1 | 8 | 1545 | |||
XM_011512472.1 | XP_011510774.1 | 8 | 1308 | |||
XM_017005789.1 | XP_016861278.1 | 7 | 1277 | |||
XM_017005790.1 | XP_016861279.1 | 8 | 1467 | |||
GenBank mRNA | AB071618.1 | - | 7 | 1478 | ||
AB079879.1 | - | 7 | 1241 | |||
AK097667.1 | - | 9 | 1531 | |||
AK122611.1 | - | 7 | 1379 | |||
AK292387.1 | - | 8 | 1507 | |||
AK296503.1 | - | 7 | 1303 | |||
AL834256.1 | - | 5 | 919 | |||
AY358141.1 | - | 7 | 1529 | |||
BC034411.1 | - | 9 | 1412 | 5172176 |