Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306215.1 | NP_001293144.1 | 2 | 846 | ||
NM_178835.4 | NP_849157.2 | 2 | 846 | |||
XM_011531634.2 | XP_011529936.1 | 2 | 893 | |||
XM_011531635.2 | XP_011529937.1 | 3 | 6380 | |||
XM_017007768.1 | XP_016863257.1 | 2 | 894 | |||
XM_017007769.1 | XP_016863258.1 | 2 | 7139 | |||
XM_017007770.1 | XP_016863259.1 | 2 | 894 | |||
XM_017007771.1 | XP_016863260.1 | 2 | 892 | |||
XM_017007772.1 | XP_016863261.1 | 2 | 892 | |||
XM_017007773.1 | XP_016863262.1 | 2 | 892 | |||
XM_017007774.1 | XP_016863263.1 | 2 | 892 | |||
XM_017007775.1 | XP_016863264.1 | 2 | 892 | |||
XM_017007776.1 | XP_016863265.1 | 2 | 894 | |||
GenBank mRNA | AK091130.1 | - | 2 | 316 | ||
AK225805.1 | - | 2 | 521 | |||
BC117407.1 | - | 2 | 357 | 40125958 | ||
BC143577.1 | - | 2 | 357 | 9052088 | ||
BM557137.1 | - | 2 | 693 | |||
DA760145.1 | - | 2 | 341 |