Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306215.1 | NP_001293144.1 | 10, 11 | 3238 | ||
NM_178835.4 | NP_849157.2 | 10, 11 | 3238 | |||
XM_011531634.2 | XP_011529936.1 | 10, 11 | 3285 | |||
XM_011531635.2 | XP_011529937.1 | 11, 12 | 8772 | |||
XM_011531645.2 | XP_011529947.1 | 6, 7 | 1023 | |||
XM_017007768.1 | XP_016863257.1 | 10, 11 | 3280 | |||
XM_017007769.1 | XP_016863258.1 | 10, 11 | 9531 | |||
XM_017007770.1 | XP_016863259.1 | 9, 10 | 3052 | |||
XM_017007771.1 | XP_016863260.1 | 10, 11 | 3284 | |||
XM_017007772.1 | XP_016863261.1 | 10, 11 | 3284 | |||
XM_017007773.1 | XP_016863262.1 | 10, 11 | 3284 | |||
XM_017007774.1 | XP_016863263.1 | 10, 11 | 3284 | |||
XM_017007775.1 | XP_016863264.1 | 10, 11 | 3284 | |||
XM_017007777.1 | XP_016863266.1 | 7, 8 | 1022 | |||
GenBank mRNA | AB074279.1 | - | 3, 4 | 358 | ||
AF450485.1 | - | 9, 10 | 1723 | |||
AK056492.1 | - | 9, 10 | 1957 | |||
AK091130.1 | - | 10, 11 | 2708 | |||
AK094136.1 | - | 5, 6 | 527 | |||
AK225805.1 | - | 10, 11 | 2913 | |||
BC117407.1 | - | 10, 11 | 2749 | 40125958 | ||
BC143577.1 | - | 10, 11 | 2749 | 9052088 |