Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278623.1 | NP_001265552.1 | 23 | 2697 | ||
NM_001278624.1 | NP_001265553.1 | 23 | 2835 | |||
NM_152995.5 | NP_694540.3 | 23 | 2737 | |||
NR_103795.1 | 24 | 2877 | ||||
GenBank mRNA | AB085695.1 | - | 23 | 2693 | ||
AB181916.1 | - | 23 | 2817 | |||
AK131285.1 | - | 21 | 2283 | |||
AK302440.1 | - | 23 | 2695 | |||
AY134856.1 | - | 7 | 741 | |||
BC018019.1 | - | 6 | 432 | |||
BC051193.1 | - | 24 | 2848 | 5271401 | ||
BU568855.1 | - | 1 | 33 |