Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040202.1 | NP_001035292.1 | 6 | 1080 | ||
GenBank mRNA | AK055774.1 | - | 6 | 868 | ||
AK126273.1 | - | 3 | 285 | |||
AK291232.1 | - | 6 | 1060 | |||
AX775925.1 | - | 6 | 1074 | |||
AY424281.1 | - | 6 | 1007 | |||
BC029604.1 | - | 5 | 848 | |||
BC031256.1 | - | 8 | 1482 | 5271599 | ||
BC047510.1 | - | 7 | 1336 | 5297045 | ||
BX648346.1 | - | 5 | 711 |