Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000972.2 | NP_000963.1 | 2 | 137 | ||
GenBank mRNA | AK222534.1 | - | 2 | 134 | ||
AK291123.1 | - | 2 | 129 | |||
AK311743.1 | - | 2 | 133 | |||
BC005128.1 | - | 1 | 120 | 3938260 | ||
BC021052.1 | - | 1 | 111 | |||
BC021979.2 | - | 1 | 109 | 4053567 | ||
BC023594.2 | - | 2 | 168 | 4841982 | ||
BC023624.2 | - | 2 | 173 | 4810336 | ||
BC032533.1 | - | 1 | 156 | |||
BC071900.1 | - | 1 | 109 | 6090366 | ||
BC071901.1 | - | 1 | 117 | 6262332 | ||
BC073802.1 | - | 1 | 108 | 4336858 | ||
BC105290.1 | - | 1 | 134 | 6495031 | ||
BX641050.1 | - | 1 | 802 | |||
M36072.1 | - | 2 | 138 | |||
X06705.1 | - | 2 | 137 |
Gene Symbol: DDIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK058145.1 | - | 8 | 1084 |