Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317791.1 | NP_001304720.1 | 2, 3 | 180 | ||
NM_144721.5 | NP_653322.1 | 2, 3 | 180 | |||
XM_005262774.3 | XP_005262831.1 | 2, 3 | 182 | |||
XM_006714109.3 | XP_006714172.1 | 2, 3 | 182 | |||
XM_011531666.2 | XP_011529968.1 | 2, 3 | 617 | |||
XM_011531667.2 | XP_011529969.1 | 2, 3 | 140 | |||
XM_017007800.1 | XP_016863289.1 | 2, 3 | 346 | |||
XM_017007801.1 | XP_016863290.1 | 2, 3 | 182 | |||
GenBank mRNA | AK303658.1 | - | 2, 3 | 140 | ||
AK313765.1 | - | 2, 3 | 564 | |||
AL833350.1 | - | 2, 3 | 140 | |||
BC022989.2 | - | 1, 2 | 94 | 5113206 | ||
BX647579.1 | - | 2, 3 | 140 | |||
DC308535.1 | - | 2, 3 | 181 |