Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317791.1 | NP_001304720.1 | 3 | 241 | ||
NM_144721.5 | NP_653322.1 | 3 | 241 | |||
NR_133920.1 | 2 | 141 | ||||
XM_005262772.2 | XP_005262829.1 | 2 | 136 | |||
XM_005262774.3 | XP_005262831.1 | 3 | 243 | |||
XM_006714109.3 | XP_006714172.1 | 3 | 243 | |||
XM_011531666.2 | XP_011529968.1 | 3 | 678 | |||
XM_011531667.2 | XP_011529969.1 | 3 | 201 | |||
XM_017007800.1 | XP_016863289.1 | 3 | 407 | |||
XM_017007801.1 | XP_016863290.1 | 3 | 243 | |||
GenBank mRNA | AK056959.1 | - | 2 | 101 | ||
AK303658.1 | - | 3 | 201 | |||
AK308914.1 | - | 2 | 101 | |||
AK309187.1 | - | 2 | 101 | |||
AK310575.1 | - | 2 | 105 | |||
AK313765.1 | - | 3 | 625 | |||
AL833350.1 | - | 3 | 201 | |||
BC022989.2 | - | 2 | 155 | 5113206 | ||
BX647579.1 | - | 3 | 201 | |||
DA995979.1 | - | 2 | 131 | |||
DC308535.1 | - | 3 | 242 |