Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013622.3 | NP_001013644.1 | 3 | 294 | ||
NM_001174070.2 | NP_001167541.1 | 3 | 341 | |||
NM_001297435.1 | NP_001284364.1 | 3 | 694 | |||
XM_005247948.3 | XP_005248005.1 | 3 | 991 | |||
XM_011513402.2 | XP_011511704.1 | 3 | 997 | |||
XM_011513403.1 | XP_011511705.1 | 3 | 991 | |||
XM_011513404.2 | XP_011511706.1 | 3 | 991 | |||
XM_011513405.1 | XP_011511707.1 | 3 | 991 | |||
XM_017007802.1 | XP_016863291.1 | 3 | 991 | |||
GenBank mRNA | AK125566.1 | - | 3 | 289 | ||
BC070112.1 | - | 3 | 294 | 5268061 | ||
HY040719.1 | - | 3 | 347 |