Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152544.2 | NP_689757.2 | 7 | 1265 | ||
XM_005247950.3 | XP_005248007.1 | 7 | 1262 | |||
XM_011513406.2 | XP_011511708.1 | 7 | 1294 | |||
XM_011513407.2 | XP_011511709.1 | 7 | 1294 | |||
XM_011513408.1 | XP_011511710.1 | 8 | 1406 | |||
XM_011513409.2 | XP_011511711.1 | 8 | 1377 | |||
GenBank mRNA | AK093044.1 | - | 3 | 271 | ||
BC035655.1 | - | 7 | 765 | 5588410 |