Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040129.2 | NP_001035218.1 | 4 | 205 | ||
GenBank mRNA | AK001520.1 | - | 5 | 674 | ||
BC128163.1 | - | 5 | 357 | 40113027 | ||
BC128164.1 | - | 4 | 205 | 40113028 | ||
DQ438947.1 | - | 3 | 143 | |||
EF108324.1 | - | 4 | 253 | |||
EF426754.1 | - | 4 | 253 |