Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 10 | 1524 | ||
NM_153223.3 | NP_694955.2 | 11 | 1605 | |||
XM_005271901.4 | XP_005271958.1 | 10 | 1386 | |||
XM_011543185.2 | XP_011541487.1 | 10 | 1939 | |||
XM_011543186.2 | XP_011541488.1 | 7 | 1218 | |||
XM_017009085.1 | XP_016864574.1 | 7 | 1083 | |||
GenBank mRNA | AK093409.1 | - | 3 | 291 | ||
AK095646.1 | - | 10 | 1526 | |||
AK296999.1 | - | 11 | 1569 | |||
BC040527.1 | - | 10 | 2188 | |||
BC144558.1 | - | 13 | 1732 | |||
CR627324.1 | - | 6 | 2761 |