Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM47E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136570.2 | NP_001130042.1 | 8 | 1187 | ||
NM_001242936.1 | NP_001229865.1 | 8 | 1260 | |||
GenBank mRNA | AK124936.1 | - | 8 | 1260 | ||
BM713546.1 | - | 4 | 432 | |||
CR627383.1 | - | 6 | 5863 |