Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBLAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203406.1 | NP_981951.1 | 2 | 1269 | ||
GenBank mRNA | AK128603.1 | - | 3 | 679 | ||
AK289778.1 | - | 2 | 821 | |||
BC038230.1 | - | 2 | 1269 | 5729339 | ||
CR749512.1 | - | 1 | 1609 |