Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144723.2 | NP_653324.1 | 3, 4 | 344 | ||
GenBank mRNA | AK055683.1 | - | 3, 4 | 257 | ||
BC056668.1 | - | 3, 4 | 261 | 4773709 | ||
BP210985.1 | - | 3, 4 | 338 | |||
BP370587.1 | - | 2, 3 | 200 |