Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006327.3 | NP_006318.1 | 4 | 475 | ||
NR_073029.1 | 4 | 475 | ||||
NR_073030.1 | 3 | 416 | ||||
XM_011539098.1 | XP_011537400.1 | 4 | 413 | |||
XM_011539099.1 | XP_011537401.1 | 4 | 413 | |||
GenBank mRNA | AF030162.1 | - | 4 | 427 | ||
AK223331.1 | - | 4 | 415 | |||
AY442341.1 | - | 4 | 427 | |||
BC062707.1 | - | 4 | 476 | 4772587 | ||
BC066951.1 | - | 4 | 425 | 5264814 | ||
BY799488.2 | - | 3 | 349 | |||
CD514420.1 | - | 4 | 351 |
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290117.1 | NP_001277046.1 | 4 | 465 | ||
NR_110767.1 | 5 | 589 | ||||
NR_135475.1 | 4 | 465 | ||||
XM_011539104.1 | XP_011537406.1 | 4 | 417 | |||
XM_017015460.1 | XP_016870949.1 | 4 | 417 | |||
XM_017015461.1 | XP_016870950.1 | 4 | 417 | |||
XM_017015462.1 | XP_016870951.1 | 4 | 417 | |||
GenBank mRNA | AK098044.1 | - | 5 | 574 | ||
AK295227.1 | - | 3 | 358 | |||
AK295370.1 | - | 4 | 417 | |||
AK316490.1 | - | 4 | 417 | |||
AK316549.1 | - | 4 | 466 | |||
AL542628.3 | - | 5 | 576 | |||
DC419977.1 | - | 3 | 407 |