Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARVELD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038603.2 | NP_001033692.2 | 2 | 1155 | ||
NM_001244734.1 | NP_001231663.1 | 2 | 1155 | |||
XM_005248445.4 | XP_005248502.1 | 2 | 1449 | |||
XM_005248446.4 | XP_005248503.1 | 2 | 1243 | |||
XM_005248447.4 | XP_005248504.1 | 2 | 1450 | |||
GenBank mRNA | AB219936.1 | - | 1 | 1081 | ||
AB219937.1 | - | 1 | 1081 | |||
AK055094.1 | - | 2 | 1151 | |||
AK292962.1 | - | 2 | 1139 | |||
BC033689.1 | - | 2 | 1140 | 5210373 | ||
DQ682656.1 | - | 2 | 1140 | |||
DQ682657.1 | - | 2 | 1140 |