Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4EBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004095.3 | NP_004086.1 | 2 | 232 | ||
GenBank mRNA | AB044548.1 | - | 2 | 205 | ||
AK312011.1 | - | 2 | 192 | |||
BC004459.1 | - | 2 | 224 | 2820216 | ||
BC058073.1 | - | 2 | 172 | 5190933 | ||
BT007162.1 | - | 2 | 160 | |||
CR456769.1 | - | 2 | 160 | |||
L36055.1 | - | 2 | 160 |